2-Chloro-3-nitrobenzonitrile
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS23611G1
AS23611G5
ACTIAS23611G1EA
56
GBP
ACTIAS23611G1
ACTIAS23611G5
2-Chloro-3-nitrobenzonitrile
2-Chloro-3-nitrobenzonitrile
Formula:
C₇H₃ClN₂O₂ MW: 182.5600 g/mol |
MDL Number:
MFCD11044838 CAS Number: 34662-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...