2-Ethoxybenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018661-25G 018661-5G
FLUO018661-25GEA 0 GBP
FLUO018661-25G FLUO018661-5G
2-Ethoxybenzonitrile
2-Ethoxybenzonitrile
Formula: C₉H₉NO
MW: 147.18 g/mol
MDL Number: MFCD00001789
CAS Number: 6609-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR