2-Ethoxybenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24962G100 AS24962G25
ACTIAS24962G100EA 158 GBP
ACTIAS24962G100 ACTIAS24962G25
2-Ethoxybenzonitrile
2-Ethoxybenzonitrile
Formula: C₉H₉NO
MW: 147.18 g/mol
MDL Number: MFCD00001789
CAS Number: 6609-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR