4-Bromo-2-tert-butoxy-1-nitrobenzene
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0869-5G
OT-0869-1G
COBBOT-0869-5GEA
0
GBP
COBBOT-0869-5G
COBBOT-0869-1G
4-Bromo-2-tert-butoxy-1-nitrobenzene
4-Bromo-2-tert-butoxy-1-nitrobenzene
Formula:
C₁₀H₁₂BrNO₃ |
MDL Number:
MFCD09909345 CAS Number: 1187386-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...