4-Bromotetrafluoroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromo-2,3,5,6-tetrafluoroaniline

010434-25G 010434-5G
FLUO010434-25GEA 0 GBP
FLUO010434-25G FLUO010434-5G
4-Bromotetrafluoroaniline
4-Bromotetrafluoroaniline
Formula: C₆H₂BrF₄N
MW: 243.9844 g/mol
MDL Number: MFCD00012144
CAS Number: 1998-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR