Methyl-2-methoxy-6-nitrobenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 2-methoxy-6-nitrobenzoate

AS21434G1 AS21434G5
ACTIAS21434G1EA 178 GBP
ACTIAS21434G1 ACTIAS21434G5
Methyl-2-methoxy-6-nitrobenzoate
Methyl 2-methoxy-6-nitrobenzoate
Formula: C₉H₉NO₅
MW: 211.17 g/mol
MDL Number: MFCD11656789
CAS Number: 77901-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR