3-Methylisonicotinonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS2689G1 AS2689G5
ACTIAS2689G1EA 97 GBP
ACTIAS2689G1 ACTIAS2689G5
3-Methylisonicotinonitrile
3-Methylisonicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile
Formula: C₇H₆N₂
MDL Number: MFCD09261048
CAS Number: 7584-05-6
EINECS: 231-491-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR