2-Amino-5-bromo-3-methylbenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4664-5G CA-4664-1G
COBBCA-4664-5GEA 0 GBP
COBBCA-4664-5G COBBCA-4664-1G
2-Amino-5-bromo-3-methylbenzoic acid
2-Amino-5-bromo-3-methylbenzoic acid
Formula: C₈H₈BrNO₂
MW: 230.0600 g/mol
MDL Number: MFCD05664860
CAS Number: 206548-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR