5-Bromo-2-hydroxybenzonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CFF852.1
005188-25G
005188-5G
CHMPCFF852.1EA
0
GBP
CHMPCFF852.1
FLUO005188-25G
FLUO005188-5G
5-Bromo-2-hydroxybenzonitrile
5-Bromo-2-hydroxybenzonitrile
Formula:
C₇H₄BrNO MW: 198.02 g/mol |
MDL Number:
MFCD00143428 CAS Number: 40530-18-5 EINECS: 254-958-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...