4-Amino-3-ethylbenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF551.5 CFF551.25 CFF551.50 005508-1G 005508-5G
CHMPCFF551.5EA 0 GBP
CHMPCFF551.5 CHMPCFF551.25 CHMPCFF551.50 FLUO005508-1G FLUO005508-5G
4-Amino-3-ethylbenzonitrile
4-Amino-3-ethylbenzonitrile
Formula: C₉H₁₀N₂
MW: 146.19 g/mol
MDL Number: MFCD00041431
CAS Number: 170230-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR