4-Amino-3,5-dibromobenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

024449-25G 024449-5G
FLUO024449-25GEA 0 GBP
FLUO024449-25G FLUO024449-5G
4-Amino-3,5-dibromobenzonitrile
4-Amino-3,5-dibromobenzonitrile
Formula: C₇H₄Br₂N₂
MW: 275.9300 g/mol
MDL Number: MFCD03094688
CAS Number: 58633-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR