4-Amino-2,5-difluorobenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007800-5G 007800-1G
FLUO007800-5GEA 0 GBP
FLUO007800-5G FLUO007800-1G
4-Amino-2,5-difluorobenzonitrile
4-Amino-2,5-difluorobenzonitrile
Formula: C₇H₄F₂N₂
MW: 154.12 g/mol
MDL Number: MFCD00190102
CAS Number: 112279-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR