2-Amino-4-fluorobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21304G25 AS21304G5
ACTIAS21304G25EA 383 GBP
ACTIAS21304G25 ACTIAS21304G5
2-Amino-4-fluorobenzonitrile
2-Amino-4-fluorobenzonitrile
Formula: C₇H₅FN₂
MW: 136.1300 g/mol
MDL Number: MFCD09955451
CAS Number: 80517-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR