3-Fluoro-5-hydroxybenzonitrile
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4583-5G
CA-4583-1G
COBBCA-4583-5GEA
0
GBP
COBBCA-4583-5G
COBBCA-4583-1G
3-Fluoro-5-hydroxybenzonitrile
3-Fluoro-5-hydroxybenzonitrile
Formula:
C₇H₄FNO MW: 137.1112 g/mol |
MDL Number:
MFCD09972133 CAS Number: 473923-95-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...