3-Phenyl-5-isoxazolecarboxylic acid
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Phenylisoxazole-5-carboxylic acid
12L-318S.500
12L-318S.1
12L-318S.5
12L-318S.10
BIOT12L-318S.500EA
0
GBP
BIOT12L-318S.500
BIOT12L-318S.1
BIOT12L-318S.5
BIOT12L-318S.10
3-Phenyl-5-isoxazolecarboxylic acid
3-Phenyl-5-isoxazolecarboxylic acid
Formula:
C₁₀H₇NO₃ MW: 189.17 g/mol |
MDL Number:
MFCD01936010 CAS Number: 14442-12-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...