6-Bromo-1H-indazole
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS2557G25
AS2557G5
ACTIAS2557G25EA
41
GBP
ACTIAS2557G25
ACTIAS2557G5
6-Bromo-1H-indazole
6-Bromo-1H-indazole
Formula:
C₇H₅BrN₂ MW: 197.0300 g/mol |
MDL Number:
MFCD03265457 CAS Number: 79762-54-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...