2,5-Dibromoisonicotinic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,5-Dibromopyridine-4-carboxylic acid

PY-1142-1G PY-1142-5G
COBBPY-1142-1GEA 0 GBP
COBBPY-1142-1G COBBPY-1142-5G
2,5-Dibromoisonicotinic acid
2,5-Dibromoisonicotinic acid
Formula: C₆H₃Br₂NO₂
MW: 280.9000 g/mol
MDL Number: MFCD08741485
CAS Number: 942473-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR