3-(4-Chlorophenyl)propiononitrile

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(4-Chlorophenyl)propanenitrile

C2931-B.1 C2931-B.5
XXXXC2931-B.1EA 0 GBP
XXXXC2931-B.1 XXXXC2931-B.5
3-(4-Chlorophenyl)propiononitrile
3-(4-Chlorophenyl)propiononitrile
Formula: C₉H₈ClN
MW: 165.6195 g/mol
MDL Number: MFCD01310802
CAS Number: 32327-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR