3-(2-Chlorophenyl)propiononitrile

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(2-Chlorophenyl)propanenitrile

C2931.5 C2931.25
XXXXC2931.5EA 0 GBP
XXXXC2931.5 XXXXC2931.25
3-(2-Chlorophenyl)propiononitrile
3-(2-Chlorophenyl)propiononitrile
Formula: C₉H₈ClN
MW: 165.6200 g/mol
MDL Number: MFCD00019873
CAS Number: 7315-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR