3-(2-Chlorophenyl)propiononitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(2-Chlorophenyl)propanenitrile

018435-1G 018435-5G
FLUO018435-1GEA 0 GBP
FLUO018435-1G FLUO018435-5G
3-(2-Chlorophenyl)propiononitrile
3-(2-Chlorophenyl)propiononitrile
Formula: C₉H₈ClN
MW: 165.6200 g/mol
MDL Number: MFCD00019873
CAS Number: 7315-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR