3-(4-Bromophenyl)propiononitrile

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(4-Bromophenyl)propanenitrile

B3200.1 B3200.5
XXXXB3200.1EA 0 GBP
XXXXB3200.1 XXXXB3200.5
3-(4-Bromophenyl)propiononitrile
3-(4-Bromophenyl)propiononitrile
Formula: C₉H₈BrN
MW: 210.0705 g/mol
MDL Number: MFCD01310795
CAS Number: 57775-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR