2-(1-Hydroxyethyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(Pyridin-2-yl)ethanol

AS21181G1 AS21181G25 AS21181G5
ACTIAS21181G1EA 19 GBP
ACTIAS21181G1 ACTIAS21181G25 ACTIAS21181G5
2-(1-Hydroxyethyl)pyridine
2-(1-Hydroxyethyl)pyridine
Formula: C₇H₉NO
MW: 123.1525 g/mol
MDL Number: MFCD06245419
CAS Number: 18728-61-5
EINECS: 242-539-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR