4-Amino-3,6-dichloropyridazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS32480G1 AS32480G5
ACTIAS32480G1EA 0 GBP
ACTIAS32480G1 ACTIAS32480G5
4-Amino-3,6-dichloropyridazine
4-Amino-3,6-dichloropyridazine
Formula: C₄H₃Cl₂N₃
MW: 163.9900 g/mol
MDL Number: MFCD01647253
CAS Number: 823-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR