2,6-Dichloroisonicotinonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Cyano-2,6-dichloropyridine

AS21572G1 AS21572G25 AS21572G5
ACTIAS21572G1EA 41 GBP
ACTIAS21572G1 ACTIAS21572G25 ACTIAS21572G5
2,6-Dichloroisonicotinonitrile
2,6-Dichloroisonicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile
Formula: C₆H₂Cl₂N₂
MW: 172.9995 g/mol
MDL Number: MFCD00173784
CAS Number: 32710-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR