3-Amino-2,5-dibromopyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,5-Dibromopyridin-3-amine

AS12619G1 AS12619G5
ACTIAS12619G1EA 0 GBP
ACTIAS12619G1 ACTIAS12619G5
3-Amino-2,5-dibromopyridine
3-Amino-2,5-dibromopyridine
Formula: C₅H₄Br₂N₂
MW: 251.9100 g/mol
MDL Number: MFCD09266224
CAS Number: 90902-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR