5-Bromo-2-fluoro-4-nitrotoluene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6965-1G HC-6965-5G
COBBHC-6965-1GEA 0 GBP
COBBHC-6965-1G COBBHC-6965-5G
5-Bromo-2-fluoro-4-nitrotoluene
5-Bromo-2-fluoro-4-nitrotoluene
Formula: C₇H₅BrFNO₂
MDL Number: MFCD11053879
CAS Number: 208165-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR