5-(1-Chloro-2-methylpropan-2-yl)-3-(4-nitrophenyl)-1,2,4-oxadiazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6524-1G HC-6524-5G
COBBHC-6524-1GEA 0 GBP
COBBHC-6524-1G COBBHC-6524-5G
5-(1-Chloro-2-methylpropan-2-yl)-3-(4-nitrophenyl)-1,2,4-oxadiazole
5-(1-Chloro-2-methylpropan-2-yl)-3-(4-nitrophenyl)-1,2,4-oxadiazole
Formula: C₁₂H₁₂ClN₃O₃
MDL Number: MFCD11855934
CAS Number: 1133116-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR