2-Hydroxy-6-methylnicotinonitrile

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile, 3-Cyano-6-methyl-2-pyridone

C3520.10 C3520.50
XXXXC3520.10EA 0 GBP
XXXXC3520.10 XXXXC3520.50
2-Hydroxy-6-methylnicotinonitrile
2-Hydroxy-6-methylnicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile
Formula: C₇H₆N₂O
MW: 134.14 g/mol
MDL Number: MFCD00006013
CAS Number: 4241-27-4
EINECS: 224-202-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR