Ethyl-2-chloro-6-methylisonicotinate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 2-chloro-6-methylpyridine-4-carboxylate, Ethyl 2-chloro-6-methylisonicotinate

AS22208G25 AS22208G5
ACTIAS22208G25EA 371 GBP
ACTIAS22208G25 ACTIAS22208G5
Ethyl-2-chloro-6-methylisonicotinate
Ethyl 2-chloro-6-methylisonicotinate
Formula: C₉H₁₀ClNO₂
MW: 199.64 g/mol
MDL Number: MFCD09842151
CAS Number: 3998-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR