5-Bromo-2-(2-hydroxyethyl)amino-3-nitropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7331-1G PY-7331-5G
COBBPY-7331-1GEA 0 GBP
COBBPY-7331-1G COBBPY-7331-5G
5-Bromo-2-(2-hydroxyethyl)amino-3-nitropyridine
5-Bromo-2-(2-hydroxyethyl)amino-3-nitropyridine
Formula: C₇H₈BrN₃O₃
MDL Number: MFCD10699705
CAS Number: 1033202-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR