2-Acetylamino-5-bromo-6-methylpyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Acetamido-5-bromo-6-methylpyridine, 6-Acetamido-3-bromo-2-methylpyridine

PY-7128-1G PY-7128-25G PY-7128-5G
COBBPY-7128-1GEA 0 GBP
COBBPY-7128-1G COBBPY-7128-25G COBBPY-7128-5G
2-Acetylamino-5-bromo-6-methylpyridine
2-Acetylamino-5-bromo-6-methylpyridine
Formula: C₈H₉BrN₂O
MW: 229.0700 g/mol
MDL Number: MFCD03909755
CAS Number: 142404-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR