Methyl-4-hydroxy-8-methoxyquinoline-2-carboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6952-5G HC-6952-1G
COBBHC-6952-5GEA 0 GBP
COBBHC-6952-5G COBBHC-6952-1G
Methyl-4-hydroxy-8-methoxyquinoline-2-carboxylate
Methyl 4-hydroxy-8-methoxyquinoline-2-carboxylate
Formula: C₁₂H₁₁NO₄
MDL Number: MFCD00100610
CAS Number: 7101-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR