Methyl-5,7-dichloro-4-hydroxyquinoline-2-carboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2266-5G HC-2266-1G
COBBHC-2266-5GEA 0 GBP
COBBHC-2266-5G COBBHC-2266-1G
Methyl-5,7-dichloro-4-hydroxyquinoline-2-carboxylate
Methyl 5,7-dichloro-4-hydroxyquinoline-2-carboxylate
Formula: C₁₁H₇Cl₂NO₃
MW: 272.842 g/mol
MDL Number: MFCD11855869
CAS Number: 130613-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR