6-Bromo-3-nitro-4-quinolinol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Bromo-3-nitroquinolin-4-ol

AS21202G1 AS21202G25 AS21202G5
ACTIAS21202G1EA 0 GBP
ACTIAS21202G1 ACTIAS21202G25 ACTIAS21202G5
6-Bromo-3-nitro-4-quinolinol
6-Bromo-3-nitro-4-quinolinol
Formula: C₉H₅BrN₂O₃
MW: 269.0516 g/mol
MDL Number: MFCD09800413
CAS Number: 853908-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR