2-Benzyloxy-3,5-difluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(Benzyloxy)-3,5-difluorophenylboronic acid

BB-5641-5G BB-5641-1G
COBBBB-5641-5GEA 0 GBP
COBBBB-5641-5G COBBBB-5641-1G
2-Benzyloxy-3,5-difluorophenylboronic acid
2-Benzyloxy-3,5-difluorophenylboronic acid
Formula: C₁₃H₁₁BF₂O₃
MDL Number: MFCD11855849
CAS Number: 1150114-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR