N-Benzyl-5-bromo-2-methoxybenzenesulphonamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0300-5G
OT-0300-1G
COBBOT-0300-5GEA
0
GBP
COBBOT-0300-5G
COBBOT-0300-1G
N-Benzyl-5-bromo-2-methoxybenzenesulphonamide
N-Benzyl 5-bromo-2-methoxybenzenesulphonamide
Formula:
C₁₄H₁₄BrNO₃S |
MDL Number:
MFCD02227932 CAS Number: 446308-82-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...