N-Methyl-3-nitrobenzenesulphonamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0107-5G
OT-0107-1G
COBBOT-0107-5GEA
0
GBP
COBBOT-0107-5G
COBBOT-0107-1G
N-Methyl-3-nitrobenzenesulphonamide
N-Methyl-3-nitrobenzenesulphonamide
Formula:
C₇H₈N₂O₄S |
MDL Number:
MFCD00474673 CAS Number: 58955-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...