Sulphamethoxypyridazine, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Amino-N-(6-methoxypyridazin-3-yl)benzenesulphonamide

M-Clarity™ quality level = MQ200

46858-250MG-REA 50.7 GBP
46858-250MG-R
Sulphamethoxypyridazine, Supelco®
Sulphamethoxypyridazine
Formula: C₁₁H₁₂N₄O₃S
MW: 280.3 g/mol
MDL Number: MFCD00057372
CAS Number: 80-35-3
EINECS: 201-272-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR