Sulphamethoxypyridazine
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EHERC16998150EA
116
GBP
EHERC16998150
Sulphamethoxypyridazine
Sulphamethoxypyridazine
Formula:
C₁₁H₁₂N₄O₃S |
MDL Number:
MFCD00057372 CAS Number: 80-35-3 EINECS: 201-272-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...