3-Fluoro-N-phenylbenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4782-5G CA-4782-25G
COBBCA-4782-5GEA 0 GBP
COBBCA-4782-5G COBBCA-4782-25G
3-Fluoro-N-phenylbenzamide
3-Fluoro-N-phenylbenzamide
Formula: C₁₃H₁₀FNO
MW: 215.223 g/mol
MDL Number: MFCD00454558
CAS Number: 1629-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR