4-Fluoro-2-nitrobenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4596-5G CA-4596-25G CA-4596-1G
COBBCA-4596-5GEA 0 GBP
COBBCA-4596-5G COBBCA-4596-25G COBBCA-4596-1G
4-Fluoro-2-nitrobenzamide
4-Fluoro-2-nitrobenzamide
Formula: C₇H₅FN₂O₃
MW: 184.1246 g/mol
MDL Number: MFCD04108565
CAS Number: 106754-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR