2-Fluoro-5-(methoxycarbamoyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-3352-5G BB-3352-1G
COBBBB-3352-5GEA 0 GBP
COBBBB-3352-5G COBBBB-3352-1G
2-Fluoro-5-(methoxycarbamoyl)phenylboronic acid
2-Fluoro-5-(methoxycarbamoyl)phenylboronic acid
Formula: C₈H₉BFNO₄
MDL Number: MFCD08436032
CAS Number: 874289-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR