3-Cyano-4-fluorophenylboronic acid ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 3-Cyano-4-fluorobenzeneboronic acid

H53041.03 H53041.MD
H53041.03EA 0 GBP
H53041.03 H53041.MD
3-Cyano-4-fluorophenylboronic acid ≥98%
3-Cyano-4-fluorophenylboronic acid

Purity: 98% CAS No.: 214210-21-6 Molecular Formula: C7H5BFNO2 Formula Weight: 164.93 Melting Point: 300° MDL No.: MFCD03095130

Formula: C₇H₅BFNO₂
MW: 164.93 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD03095130
CAS Number: 214210-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR