4-Methoxy-N-tert-butylbenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-3279-5G HC-3279-25G
COBBHC-3279-5GEA 0 GBP
COBBHC-3279-5G COBBHC-3279-25G
4-Methoxy-N-tert-butylbenzamide
4-Methoxy-N-tert-butylbenzamide
Formula: C₁₂H₁₇NO₂
MDL Number: MFCD00751505
CAS Number: 19486-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR