3-Methoxy-N-tert-butylbenzamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-1149-5G
OT-1149-25G
OT-1149-1G
COBBOT-1149-5GEA
0
GBP
COBBOT-1149-5G
COBBOT-1149-25G
COBBOT-1149-1G
3-Methoxy-N-tert-butylbenzamide
3-Methoxy-N-tert-butylbenzamide
Formula:
C₁₂H₁₇NO₂ |
MDL Number:
MFCD00157792 CAS Number: 49834-28-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...