2-Methoxy-N-tert-butylbenzamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4792-25G
CA-4792-1G
CA-4792-5G
COBBCA-4792-25GEA
0
GBP
COBBCA-4792-25G
COBBCA-4792-1G
COBBCA-4792-5G
2-Methoxy-N-tert-butylbenzamide
2-Methoxy-N-tert-butylbenzamide
Formula:
C₁₂H₁₇NO₂ |
MDL Number:
MFCD00751503 CAS Number: 88105-14-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...