4-Chloro-N,N-dimethylbenzamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
N,N-Dimethyl 4-chlorobenzamide
CA-4544-5G
CA-4544-25G
CA-4544-1G
COBBCA-4544-5GEA
0
GBP
COBBCA-4544-5G
COBBCA-4544-25G
COBBCA-4544-1G
4-Chloro-N,N-dimethylbenzamide
4-Chloro-N,N-dimethylbenzamide
Formula:
C₉H₁₀ClNO MW: 183.6400 g/mol |
MDL Number:
MFCD00463859 CAS Number: 14062-80-7 EINECS: 237-909-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...