4-Chloro-3-(ethylcarbamoyl)phenylboronic acid ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Chloro-3-(ethylcarbamoyl)benzeneboronic acid

H53175.MDEA 0 GBP
H53175.MD
4-Chloro-3-(ethylcarbamoyl)phenylboronic acid ≥98%
4-Chloro-3-(ethylcarbamoyl)phenylboronic acid
Formula: C₉H₁₁BClNO₃
MW: 227.45 g/mol
Melting Pt: 220…224 °C
Storage Temperature: Ambient
MDL Number: MFCD07783861
CAS Number: 871332-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR