3-Chloro-4-(ethylcarbamoyl)phenylboronic acid ≥97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: (3-Chloro-4-(ethylcarbamoyl)phenyl)boronic acid3-Chloro-4-(ethylcarbamoyl)benzeneboronic acid

H53162.MDEA 0 GBP
H53162.MD
3-Chloro-4-(ethylcarbamoyl)phenylboronic acid ≥97%
3-Chloro-4-(ethylcarbamoyl)phenylboronic acid
Formula: C₉H₁₁BClNO₃
MW: 227.45 g/mol
Melting Pt: 142…146 °C
Storage Temperature: Ambient
MDL Number: MFCD07363767
CAS Number: 850589-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR