4-Chlorobenzonitrile-2,5-diboronic acid neopentyl glycol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-8590-1G BB-8590-250M
COBBBB-8590-1GEA 0 GBP
COBBBB-8590-1G COBBBB-8590-250M
4-Chlorobenzonitrile-2,5-diboronic acid neopentyl glycol ester
4-Chlorobenzonitrile-2,5-diboronic acid neopentyl glycol ester
Formula: C₁₇H₂₂B₂ClNO₄
MDL Number: MFCD09878542
CAS Number: 1072944-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR